Methylrød, Supelco®
Glass bottle, 100 g
: Merck
: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
- :493-52-7
- :C₁₅H₁₅N₃O₂
- :MFCD00002425
- :269.3 g/mol
- :12,06199
- :178…182 °C
- :1,31 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- :Stue temperatur
- :207-776-1
- Solubility:(20 °C) slightly soluble
- Melting Point:178 - 182 °C
- Molar Mass:269.31 g/mol
- Bulk Density:300 - 500 kg/m3
- Hazard Class:9
- Hazard Class Description:Miscellaneous Hazardous Material
- Pakning:Glass bottle
- Pakn:100 g



