Methylrød, Supelco®
Glass bottle, 25 g
Leverandør: Merck
Gå til:
Product Details & Documents
Synonymer: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
- CAS nummer:493-52-7
- Formel:C₁₅H₁₅N₃O₂
- MDL nummer:MFCD00002425
- Molecular Weight:269.3 g/mol
- Merck Index:12,06199
- Smeltende Pt:178…182 °C
- Tæthed:1,31 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- Opbevaringstemperatur:Stue temperatur
- EINECS:207-776-1
Specifikationer
Specifikationstestresultater
- Solubility:(20 °C) slightly soluble
- Melting Point:178 - 182 °C
- Molar Mass:269.31 g/mol
- Bulk Density:300 - 500 kg/m3
- Hazard Class:9
- Hazard Class Description:Miscellaneous Hazardous Material
Specifikationer
- Pakning:Glass bottle
- Pakn:25 g



