Colchicin
Glass bottle for solids, 250 mg
Catalog # SERA77120.01
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: 7α-H-Colchicine, (S)-(-)-Colchicine
- CAS Number:64-86-8
- Formula:C₂₂H₂₅NO₆
- MDL Number:MFCD00078484
- Molecular Weight:399.44 g/mol
- Merck Index:12,02536
- Melting Pt:154…156 °C
- Density:1,32 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:200-598-5
- UN:1544
- ADR:6.1,I
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:250 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



