Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Lecithin (from soy bean)

Glass bottle for solids, 1 kg
Katalog # SERA57556.02
Marketsource
Leverandør:   Serva
Lecithin (from soy bean)
Katalog # SERA57556.02
Leverandør:  Serva
Marketsource
CAS nummer:  8030-76-0
Lecithin (from soy bean)

Glass bottle for solids, 1 kg

CAS nummer:  8030-76-0
Price
1.015,60 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
  • CAS nummer:8030-76-0
  • MDL nummer:MFCD00147406
  • EINECS:310-129-7

 

Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg

Specifikationer