Pepstatin A
Glass bottle for solids, 100 mg
Katalog # SERA52682.04
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 mg
- Cat. No.:SERA52682.04
- CAS-nr:26305-03-3
- Molecular formula:C₃₄H₆₃N₅O₉
- Protein/peptide name:Pepstatin A
- MDL No.:MFCD00060740
- Opbevaring betingelser:Refrigerator
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



