Pepstatin A
Glass bottle for solids, 25 mg
Katalog # SERA52682.03
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:25 mg
- Cat. No.:SERA52682.03
- CAS-nr:26305-03-3
- Molecular formula:C₃₄H₆₃N₅O₉
- Protein/peptide name:Pepstatin A
- MDL No.:MFCD00060740
- Opbevaring betingelser:Refrigerator



