Orders LinkContactUsLinkComponent
Base materials
Base materials
Peptone from casein (tryptic digest), Glass bottle for solids, 250 g
Katalog # SERA48600.04
Marketsource
Leverandør:   Serva
Base materials
Katalog # SERA48600.04
Leverandør:  Serva
Marketsource
CAS nummer:  
Base materials
Peptone from casein (tryptic digest), Glass bottle for solids, 250 g
CAS nummer:  
Price
358,45 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents
High quality source of peptides and amino acids produced by enzymatic digestion.
  • Suitable as nutrient for laboratory media and industrial fermentation
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g
  • Beskrivelse:
    Peptone from casein (tryptic digest)