Kaliumchlorid, EMSURE® for analysis, Supelco®
Plastic bottle, 5 kg
: Merck
: Potassiumchloride
- :7447-40-7
- :KCl
- :MFCD00011360
- :74.55 g/mol
- :14,07621
- :1420 °C (1013 hPa)
- :773 °C
- :1,98 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ300
- :Stue temperatur
- :231-211-8
- Solubility:347 g/l (20 °C)
- Melting Point:773 °C
- Molar Mass:74.55 g/mol
- Bulk Density:1000 kg/m3
- Boiling Point:1413 °C (1013 hPa)
- Density:1.98 g/cm3 (20 °C)
- pH:5.5 - 8.0 (50 g/l, H2O, 25 °C)
- Pakning:Plastic bottle
- Pakn:5 kg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



