D(+)-Xylose
Glass bottle for solids, 500 g
Katalog # SERA38530.02
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
- CAS nummer:58-86-6
- Formel:C₅H₁₀O₅
- MDL nummer:MFCD00151475
- Molecular Weight:150.13 g/mol
- Merck Index:13,10142
- Kogende Pt:331 °C (1013 hPa)
- Smeltende Pt:154 °C
- Tæthed:1,525 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-400-7
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 g



