Orders LinkContactUsLinkComponent
L(+)-Valin
Glass bottle for solids, 500 g
  SERA38064.03
Marketsource
 :   Serva
L(+)-Valin
  SERA38064.03
 :  Serva
Marketsource
 :  72-18-4
L(+)-Valin
Glass bottle for solids, 500 g
 :  72-18-4
1 * 500 g
 :  H-Val-OH, Valine, (S)-(+)-Valine
  •  :72-18-4
  •  :C₅H₁₁NO₂
  •  :MFCD00064220
  •  :117.15 g/mol
  •  :13,09458
  •  :217 °C (1013 hPa)
  •  :86 °C
  •  :1,284 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :200-773-6
  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g