Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

L(-)-Tryptophan

Glass bottle for solids, 500 g
Katalog # SERA37422.04
Marketsource
Leverandør:   Serva
L(-)-Tryptophan
Katalog # SERA37422.04
Leverandør:  Serva
Marketsource
CAS nummer:  73-22-3
L(-)-Tryptophan

Glass bottle for solids, 500 g

CAS nummer:  73-22-3
Price
2.531,53 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Synonymer:  L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
  • CAS nummer:73-22-3
  • Formel:C₁₁H₁₂N₂O₂
  • MDL nummer:MFCD00064340
  • Molecular Weight:204.23 g/mol
  • Merck Index:12,09929
  • Smeltende Pt:290 °C
  • Tæthed:1,34 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-795-6
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer