Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
n-Hexan, LiChrosolv® for liquid chromatography, Supelco®

n-Hexan, LiChrosolv® for liquid chromatography, Supelco®

Stainless steel barrel, returnable, 30 L
Katalog # 1.04391.9030
Brandfarlig
Sundhed
Irriterende
Miljø
Leverandør:   Merck
n-Hexan, LiChrosolv® for liquid chromatography, Supelco®
Katalog # 1.04391.9030
Leverandør:  Merck
CAS nummer:  110-54-3
n-Hexan, LiChrosolv® for liquid chromatography, Supelco®
n-Hexan, LiChrosolv® for liquid chromatography, Supelco®

Stainless steel barrel, returnable, 30 L

CAS nummer:  110-54-3
Price
Spørg for pris
Log ind for tilgængelighed
1 * 30 L
Product Details & Documents

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in 'gradient grade' as well as 'isocratic grade'. This minimises the gradient effect of the solvent involved.

Synonymer:  Hexyl hydride, Hexanen, Hexane
  • CAS nummer:110-54-3
  • Formel:H₃C(CH₂)₄CH₃
  • MDL nummer:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:13,04712
  • Kogende Pt:69 °C (1013 hPa)
  • Smeltende Pt:–94,3 °C
  • Tæthed:0,659 g/cm³ (20 °C)
  • Flash Pt:–22 °C (closed cup)
  • Opbevaringstemperatur:Stue temperatur
  • FN:1208
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:203-777-6
Specifikationer

Specifikationer

  • Pakning:
    Stainless steel barrel, returnable
  • Pakn:
    30 L

Specifikationer