Tricine
Glass bottle for solids, 500 g
Catalog # SERA37195.03
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: 2-((1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl)amino)acetic acid, Tricine buffer, N-(Tris(hydroxymethyl)methyl)glycine
- CAS Number:5704-04-1
- Formula:(HOCH₂)₃CNHCH₂CO₂H
- MDL Number:MFCD00004277
- Molecular Weight:179.17 g/mol
- Merck Index:13,09724
- Melting Pt:187 °C
- Density:1,523 g/cm³ (19.85 °C)
- Storage Temperature:Stue temperatur
- EINECS:227-193-6
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



