Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

n-Hexan, Uvasol® for spectroscopy, Supelco®

Glass bottle, 2,5 L
Katalog # 1.04372.2500
Brandfarlig
Sundhed
Irriterende
Miljø
Leverandør:   Merck
n-Hexan, Uvasol® for spectroscopy, Supelco®
Katalog # 1.04372.2500
Leverandør:  Merck
CAS nummer:  110-54-3
n-Hexan, Uvasol® for spectroscopy, Supelco®
n-Hexan, Uvasol® for spectroscopy, Supelco®

Glass bottle, 2,5 L

CAS nummer:  110-54-3
Price
2.570,00 kr
Log ind for tilgængelighed
1 * 2,5 L
Product Details & Documents
Synonymer:  Hexyl hydride, Hexanen, Hexane
  • CAS nummer:110-54-3
  • Formel:H₃C(CH₂)₄CH₃
  • MDL nummer:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:13,04712
  • Kogende Pt:69 °C (1013 hPa)
  • Smeltende Pt:–94,3 °C
  • Tæthed:0,659 g/cm³ (20 °C)
  • Flash Pt:–22 °C
  • Opbevaringstemperatur:Stue temperatur
  • FN:1208
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:203-777-6
Specifikationer

Specifikationstestresultater

  • Purity (GC):
    ≥99.0%
  • Evaporation Residue:
    ≤0.0002%
  • Water:
    ≤0.005%
  • Color:
    ≤10 Hazen
  • Acidity:
    ≤0.0002 meq/g
  • Alkalinity:
    ≤0.0002 meq/g
  • Fluorescence (as quinine at 254 nm):
    ≤1.0 ppb
  • Fluorescence (as quinine at 365 nm):
    ≤1.0 ppb
  • Transmission (at 195 nm):
    ≥10%
  • Transmission (at 210 nm):
    ≥60%
  • Transmission (at 217 nm):
    ≥80%
  • Transmission (at 225 nm):
    ≥90%
  • Transmission (from 245 nm):
    ≥98%
  • Absorbance (at 195 nm):
    ≤1.00
  • Absorbance (at 210 nm):
    ≤0.22
  • Absorbance (at 217 nm):
    ≤0.10
  • Absorbance (at 225 nm):
    ≤0.05
  • Absorbance (from 245 nm):
    ≤0.01

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    2,5 L

Specifikationer