n-Hexan, SupraSolv® for GC-ECD/FID, Supelco®
Glass bottle, 2,5 L
Leverandør: Merck
n-Hexan, SupraSolv® for GC-ECD/FID, Supelco®
Katalog # 1.04371.2500
Leverandør: Merck
CAS nummer: 110-54-3
Gå til:
Product Details & Documents
SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.
Synonymer: Hexyl hydride, Hexanen, Hexane
- CAS nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL nummer:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Kogende Pt:69 °C (1013 hPa)
- Smeltende Pt:–94,3 °C
- Tæthed:0,659 g/cm³ (20 °C)
- Flash Pt:–22 °C
- Opbevaringstemperatur:Stue temperatur
- FN:1208
- ADR:3,II
- :M-Clarity™ quality level = MQ100
- EINECS:203-777-6
Documents
Certifikater
Søg efter partinummerMERC1.04371.2500_TDS_EN ProductPage_104371
Produktspecifikation
Merck Individual products and tailor-made chemicals for your technical applications Merck SupraSolv® High Purity Solvents for Gas Chromatography
Produktlitteratur
msds_104371
SDS
Specifikationer
Specifikationstestresultater
- Purity (GC):≥98.0 %
- Sum of hexane isomers + methylcyclopentane (GC):≥99.8 %
- Identity (IR):conforms
- Evaporation residue:≤3.0 mg/l
- Water:≤0.01 %
- Color:≤ 10 Hazen
- GC/ECD (retention range 1,2,4-trichlorobenzene to decachlorobiphenyle, individual signals (lindane standard)):≤ 3 pg/ml
- GC/FID (retention range n-undecane to n-tetracontane,individual signals (n-tetradecane standard)):≤ 3 ng/ml
- suitable for residue analysis:0.66 g/cm3 (20 °C)
- pH:(H2O) not applicable
- Ignition Point:240 °C
- Viscosity kinematic:0.50 mm2/s (20 °C)
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



