n-Hexan, SupraSolv® for GC-ECD/FID, Supelco®
Glass bottle, 1 L
Supplier: Merck
n-Hexan, SupraSolv® for GC-ECD/FID, Supelco®
Catalog # 1.04371.1000
Supplier: Merck
CAS Number: 110-54-3
Jump to:
Product Details & Documents
SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.
Synonyms: Hexyl hydride, Hexanen, Hexane
- CAS Number:110-54-3
- Formula:H₃C(CH₂)₄CH₃
- MDL Number:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Boiling Pt:69 °C (1013 hPa)
- Melting Pt:–94,3 °C
- Density:0,659 g/cm³ (20 °C)
- Flash Pt:–22 °C
- Storage Temperature:Stue temperatur
- UN:1208
- ADR:3,II
- :M-Clarity™ quality level = MQ100
- EINECS:203-777-6
Specifications
Specification Test Results
- Purity (GC):≥98.0 %
- Sum of hexane isomers + methylcyclopentane (GC):≥99.8 %
- Identity (IR):conforms
- Evaporation residue:≤3.0 mg/l
- Water:≤0.01 %
- Color:≤ 10 Hazen
- GC/ECD (retention range 1,2,4-trichlorobenzene to decachlorobiphenyle, individual signals (lindane standard)):≤ 3 pg/ml
- GC/FID (retention range n-undecane to n-tetracontane,individual signals (n-tetradecane standard)):≤ 3 ng/ml
- suitable for residue analysis:0.66 g/cm3 (20 °C)
- pH:(H2O) not applicable
- Ignition Point:240 °C
- Viscosity kinematic:0.50 mm2/s (20 °C)
Specifications
- Pakning:Glass bottle
- Pakn:1 L



