n-Hexan, EMSURE® ACS for analysis, Supelco®
Glass bottle, 2,5 L
Leverandør: Merck
n-Hexan, EMSURE® ACS for analysis, Supelco®
Katalog # 1.04367.2500
Leverandør: Merck
CAS nummer: 110-54-3
Gå til:
Product Details & Documents
EMSURE® grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE® provides worldwide best and most extensive product specifications. We declare our EMSURE® range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.
Synonymer: Hexyl hydride, Hexanen, Hexane
- CAS nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL nummer:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Kogende Pt:69 °C (1013 hPa)
- Smeltende Pt:–94,3 °C
- Tæthed:0,659 g/cm³ (20 °C)
- Flash Pt:–22 °C
- Opbevaringstemperatur:Stue temperatur
- FN:1208
- ADR:3,II
- :M-Clarity™ quality level = MQ300
- EINECS:203-777-6
Specifikationer
Specifikationstestresultater
- Purity (GC):≥ 99.0 %
- Identity (IR):conforms
- Color:≤ 10 Hazen
- Alkalinity:≤ 0.0002 meq/g
- Acidity:≤ 0.0002 meq/g
- Density (d 20 °C/ 4 °C):0.659 - 0.662
- Thiophene:conforms
- Aromatics (as benzene):≤ 0.01 %
- Sulfur compounds (as S):≤ 0.005 %
- Matter discolored by H₂SO₄:≤ 10 Hazen
- Al (Aluminium):≤ 0.00005 %
- B (Boron):≤ 0.000002 %
- Ba (Barium):≤ 0.00001 %
- Ca (Calcium):≤ 0.00005 %
- Cd (Cadmium):≤ 0.000005 %
- Co (Cobalt):≤ 0.000002 %
- Cr (Chromium):≤ 0.000002 %
- Cu (Copper):≤ 0.000002 %
- Fe (Iron):≤ 0.00001 %
- Mg (Magnesium):≤ 0.00001 %
- Mn (Manganese):≤ 0.000002 %
- Ni (Nickel):≤ 0.000002 %
- Pb (Lead):≤ 0.00001 %
- Sn (Tin):≤ 0.00001 %
- Zn (Zinc):≤ 0.00001 %
- Evaporation residue:≤ 0.001 %
- Water:≤ 0.005 %
Specifikationer
- Pakning:Glass bottle
- Pakn:2,5 L



