Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
N,N,N',N'-Tetramethylethylendiamin

N,N,N',N'-Tetramethylethylendiamin

Glass bottle, 100 mL
Catalog # SERA35925.01
Flammable
Corrosive
Irritant
Marketsource
Supplier:   Serva
N,N,N',N'-Tetramethylethylendiamin
Catalog # SERA35925.01
Supplier:  Serva
Marketsource
CAS Number:  110-18-9
N,N,N',N'-Tetramethylethylendiamin
N,N,N',N'-Tetramethylethylendiamin

Glass bottle, 100 mL

CAS Number:  110-18-9
Price
186,69 kr
Log in for availability
1 * 100 mL
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Synonyms:  1,2-Bis(dimethylamino)ethane, TMEDA, N,N,N',N'-Tetramethylethylenediamine
  • CAS Number:110-18-9
  • Formula:C₆H₁₆N₂
  • MDL Number:MFCD00008335
  • Molecular Weight:116.21 g/mol
  • Boiling Pt:120 °C (1013 hPa)
  • Melting Pt:–64 °C
  • Density:0,7765 g/cm³ (20 °C)
  • Flash Pt:17 °C (closed cup)
  • Storage Temperature:Stue temperatur
  • EINECS:203-744-6
  • UN:2372
  • ADR:3,II
Specifications

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    100 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.