Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Streptomycin sulfat

Streptomycin sulfat

Glass bottle for solids, 10 g
Katalog # SERA35500.01
Sundhed
Irriterende
Marketsource
Leverandør:   Serva
Streptomycin sulfat
Katalog # SERA35500.01
Leverandør:  Serva
Marketsource
CAS nummer:  3810-74-0
Streptomycin sulfat

Glass bottle for solids, 10 g

CAS nummer:  3810-74-0
Price
171,76 kr
Log ind for tilgængelighed
1 * 10 g
Product Details & Documents
  • CAS nummer:3810-74-0
  • Formel:C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
  • MDL nummer:MFCD00037023
  • Molecular Weight:1457.4 g/mol
  • Merck Index:12,08983
  • Smeltende Pt:250 °C
  • Opbevaringstemperatur:Køleskab
  • EINECS:223-286-0
  • FN:2811
  • ADR:6.1,III

Documents

Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    10 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.