Sølvnitrat
Plastic bottle for solids, 500 g
Catalog # SERA35110.03
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
- CAS Number:7761-88-8
- Formula:AgNO₃
- MDL Number:MFCD00003414
- Molecular Weight:169.87 g/mol
- Merck Index:12,08661
- Boiling Pt:444 °C (1013 hPa)
- Melting Pt:210 °C
- Density:4,35 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:231-853-9
- UN:1493
- ADR:5.1,II
- REACH:01-2119513705-43
Specifications
Specifications
- Pakning:Plastic bottle for solids
- Pakn:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



