Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
D(+)-Glucose monohydrat, Millipore®

D(+)-Glucose monohydrat, Millipore®

Plastic bottle, 1 kg
Katalog # 1.04074.1000
Leverandør:   Merck
D(+)-Glucose monohydrat, Millipore®
Katalog # 1.04074.1000
Leverandør:  Merck
CAS nummer:  14431-43-7
D(+)-Glucose monohydrat, Millipore®
D(+)-Glucose monohydrat, Millipore®

Plastic bottle, 1 kg

CAS nummer:  14431-43-7
Price
447,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Synonymer:  (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate
  • CAS nummer:14431-43-7
  • Formel:C₆H₁₂O₆·H₂O
  • MDL nummer:MFCD00149450
  • Molecular Weight:198.17 g/mol
  • Smeltende Pt:83 °C
  •  :M-Clarity™ quality level = MQ100
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-075-1

Documents

Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle
  • Pakn:
    1 kg

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.