Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Saponin

Saponin

Glass bottle for solids, 250 g
Catalog # SERA34655.02
Marketsource
Supplier:   Serva
Saponin
Catalog # SERA34655.02
Supplier:  Serva
Marketsource
CAS Number:  8047-15-2
Saponin

Glass bottle for solids, 250 g

CAS Number:  8047-15-2
Price
1.150,02 kr
Log in for availability
1 * 250 g
Product Details & Documents
  • CAS Number:8047-15-2
  • MDL Number:MFCD00081981
  • Density:1,015…1.02 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:232-462-6
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g

Specifications