Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Saponin

Saponin

Glass bottle for solids, 50 g
Katalog # SERA34655.01
Marketsource
Leverandør:   Serva
Saponin
Katalog # SERA34655.01
Leverandør:  Serva
Marketsource
CAS nummer:  8047-15-2
Saponin

Glass bottle for solids, 50 g

CAS nummer:  8047-15-2
Price
276,30 kr
Log ind for tilgængelighed
1 * 50 g
Product Details & Documents
  • CAS nummer:8047-15-2
  • MDL nummer:MFCD00081981
  • Tæthed:1,015…1.02 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:232-462-6
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    50 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.