Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Fuchsin (syre)

Fuchsin (syre)

Glass bottle for solids, 25 g
Catalog # SERA34597.01
Marketsource
Supplier:   Serva
Fuchsin (syre)
Catalog # SERA34597.01
Supplier:  Serva
Marketsource
CAS Number:  3244-88-0
Fuchsin (syre)

Glass bottle for solids, 25 g

CAS Number:  3244-88-0
Price
388,32 kr
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  CI 42685
  • CAS Number:3244-88-0
  • Formula:C₂₀H₁₇N₃Na₂O₉S₃
  • MDL Number:MFCD00013286
  • Molecular Weight:585.55 g/mol
  • Merck Index:12,00109
  • Melting Pt:> 130 °C
  • Storage Temperature:Stue temperatur
  • EINECS:221-816-5
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.