Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Rifampicin

Rifampicin

Glass bottle for solids, 5 g
Katalog # SERA34514.02
Marketsource
Leverandør:   Serva
Rifampicin
Katalog # SERA34514.02
Leverandør:  Serva
Marketsource
CAS nummer:  13292-46-1
Rifampicin

Glass bottle for solids, 5 g

CAS nummer:  13292-46-1
Price
1.426,32 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  Rifampin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifamycin AMP
  • CAS nummer:13292-46-1
  • Formel:C₄₃H₅₈N₄O₁₂
  • MDL nummer:MFCD00151389
  • Molecular Weight:822.95 g/mol
  • Merck Index:12,08382
  • Smeltende Pt:183…188 °C
  • EINECS:236-312-0
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.