Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Rifampicin

Glass bottle for solids, 1 g
Katalog # SERA34514.01
Marketsource
Leverandør:   Serva
Rifampicin
Katalog # SERA34514.01
Leverandør:  Serva
Marketsource
CAS nummer:  13292-46-1
Rifampicin

Glass bottle for solids, 1 g

CAS nummer:  13292-46-1
Price
388,32 kr
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents
Synonymer:  Rifampin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifamycin AMP
  • CAS nummer:13292-46-1
  • Formel:C₄₃H₅₈N₄O₁₂
  • MDL nummer:MFCD00151389
  • Molecular Weight:822.95 g/mol
  • Merck Index:12,08382
  • Smeltende Pt:183…188 °C
  • EINECS:236-312-0
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 g

Specifikationer