Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ponceau S

Ponceau S

Glass bottle for solids, 25 g
Catalog # SERA33429.02
Irritant
Marketsource
Supplier:   Serva
Ponceau S
Catalog # SERA33429.02
Supplier:  Serva
Marketsource
CAS Number:  6226-79-5
Ponceau S

Glass bottle for solids, 25 g

CAS Number:  6226-79-5
Price
336,04 kr
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt
  • CAS Number:6226-79-5
  • Formula:C₂₂H₁₂N₄Na₄O₁₃S₄
  • MDL Number:MFCD00003892
  • Molecular Weight:760.56 g/mol
  • Storage Temperature:Stue temperatur
  • EINECS:228-319-2
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.