Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Ponceau S

Ponceau S

Glass bottle for solids, 5 g
Katalog # SERA33429.01
Irriterende
Marketsource
Leverandør:   Serva
Ponceau S
Katalog # SERA33429.01
Leverandør:  Serva
Marketsource
CAS nummer:  6226-79-5
Ponceau S

Glass bottle for solids, 5 g

CAS nummer:  6226-79-5
Price
186,69 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  3-Hydroxy-4-(2-sulpho-4-[4-sulphophenylazo]phenylazo)-2,7-naphthalenedisulphonic acid sodium salt, CI 27195, Ponceau, 3-Hydroxy-4-((2-sulpho-4-((4-sulphophenyl)diazenyl)phenyl)diazenyl)naphthalene-2,7-disulphonic acid, tetrasodium salt
  • CAS nummer:6226-79-5
  • Formel:C₂₂H₁₂N₄Na₄O₁₃S₄
  • MDL nummer:MFCD00003892
  • Molecular Weight:760.56 g/mol
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:228-319-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.