Polyethylenglycol 40.000
Glass bottle for solids, 500 g
SERA33139.01
Marketsource
: Serva
- :25322-68-3
- :H(OCH₂CH₂)nOH
- :40000 g/mol
- :Stue temperatur
- :500-038-2
- Pakning:Glass bottle for solids
- Pakn:500 g