Orders LinkContactUsLinkComponent
Phenol rød
Glass bottle for solids, 5 g
  SERA32095.01
Marketsource
 :   Serva
Phenol rød
  SERA32095.01
 :  Serva
Marketsource
 :  143-74-8
Phenol rød
Glass bottle for solids, 5 g
 :  143-74-8
141,88 kr
1 * 5 g
 :  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  •  :143-74-8
  •  :C₁₉H₁₄O₅S
  •  :MFCD00003552
  •  :354.38 g/mol
  •  :13,07329
  •  :> 300 °C
  •  :Stue temperatur
  •  :205-609-7
  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g