Phenol rød
Glass bottle for solids, 5 g
SERA32095.01
Marketsource
: Serva
: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
- :143-74-8
- :C₁₉H₁₄O₅S
- :MFCD00003552
- :354.38 g/mol
- :13,07329
- :> 300 °C
- :Stue temperatur
- :205-609-7
- Pakning:Glass bottle for solids
- Pakn:5 g