Estradiol
Glass bottle for solids, 5 g
Katalog # SERA31100.02
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: beta-Estradiol, 17β-Estradiol
- CAS nummer:50-28-2
- Formel:C₁₈H₂₄O₂
- MDL nummer:MFCD00003693
- Molecular Weight:272.39 g/mol
- EINECS:200-023-8
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:5 g