Ninhydrin
Glass bottle for solids, 25 g
Catalog # SERA30410.01
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
- CAS Number:485-47-2
- Formula:C₉H₆O₄
- MDL Number:MFCD00003791
- Molecular Weight:178.14 g/mol
- Merck Index:13,06582
- Boiling Pt:351 °C (1013 hPa)
- Melting Pt:250…258 °C
- Density:0,862 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:207-618-1
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:25 g



