Orders LinkContactUsLinkComponent
Cyclohexan, EMPLURA®, Supelco®
Cyclohexan, EMPLURA®, Supelco®
Glass bottle, 2,5 L
Catalog # 1.02832.2500
Flammable
Health
Irritant
Environment
Supplier:   Merck
Cyclohexan, EMPLURA®, Supelco®
Catalog # 1.02832.2500
Supplier:  Merck
CAS Number:  110-82-7
Cyclohexan, EMPLURA®, Supelco®
Cyclohexan, EMPLURA®, Supelco®
Glass bottle, 2,5 L
CAS Number:  110-82-7
Price
624,00 kr
Log in for availability
1 * 2,5 L
Product Details & Documents
EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA® we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA® solvents provide adequate specifications with the most important parameters.
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (1013 hPa)
  • Melting Pt:7 °C
  • Density:0,7785 g/cm³ (20 °C)
  • Flash Pt:–18 °C
  • Storage Temperature:Stue temperatur
  • UN:1145
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ200
  • EINECS:203-806-2
Documents
Certificates
Search by Lot Number
Specifications
Specifications
  • Pakning:
    Glass bottle
  • Pakn:
    2,5 L
Specifications