Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cyclohexan, LiChrosolv® for HPLC, Supelco®

Cyclohexan, LiChrosolv® for HPLC, Supelco®

Glass bottle, 2,5 L
Katalog # 1.02827.2500
Brandfarlig
Sundhed
Irriterende
Miljø
Leverandør:   Merck
Cyclohexan, LiChrosolv® for HPLC, Supelco®
Katalog # 1.02827.2500
Leverandør:  Merck
CAS nummer:  110-82-7
Cyclohexan, LiChrosolv® for HPLC, Supelco®
Cyclohexan, LiChrosolv® for HPLC, Supelco®

Glass bottle, 2,5 L

CAS nummer:  110-82-7
Price
3.290,00 kr
Log ind for tilgængelighed
1 * 2,5 L
Product Details & Documents
Synonymer:  Hexahydrobenzene, Hexamethylene
  • CAS nummer:110-82-7
  • Formel:C₆H₁₂
  • MDL nummer:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Kogende Pt:81 °C (1013 hPa)
  • Smeltende Pt:7 °C
  • Tæthed:0,7785 g/cm³ (20 °C)
  • Flash Pt:–18 °C
  • Opbevaringstemperatur:Stue temperatur
  • FN:1145
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:203-806-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    2,5 L

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.