Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cyclohexan, LiChrosolv® for HPLC, Supelco®

Cyclohexan, LiChrosolv® for HPLC, Supelco®

Glass bottle, 1 L
Katalog # 1.02827.1000
Brandfarlig
Sundhed
Irriterende
Miljø
Leverandør:   Merck
Cyclohexan, LiChrosolv® for HPLC, Supelco®
Katalog # 1.02827.1000
Leverandør:  Merck
CAS nummer:  110-82-7
Cyclohexan, LiChrosolv® for HPLC, Supelco®
Cyclohexan, LiChrosolv® for HPLC, Supelco®

Glass bottle, 1 L

CAS nummer:  110-82-7
Price
1.570,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
Synonymer:  Hexahydrobenzene, Hexamethylene
  • CAS nummer:110-82-7
  • Formel:C₆H₁₂
  • MDL nummer:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Kogende Pt:81 °C (1013 hPa)
  • Smeltende Pt:7 °C
  • Tæthed:0,7785 g/cm³ (20 °C)
  • Flash Pt:–18 °C
  • Opbevaringstemperatur:Stue temperatur
  • FN:1145
  • ADR:3,II
  •  :M-Clarity™ quality level = MQ100
  • EINECS:203-806-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

Specifikationer