MOPS
Glass bottle for solids, 500 g
SERA29836.03
Marketsource
: Serva
: 3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
- :1132-61-2
- :C₇H₁₅NO₄S
- :MFCD00006183
- :209.27 g/mol
- :13,06288
- :> 250 °C
- :110 °C (closed cup)
- :Stue temperatur
- :214-478-5
- Pakning:Glass bottle for solids
- Pakn:500 g