Orders LinkContactUsLinkComponent
MOPS
Glass bottle for solids, 500 g
  SERA29836.03
Marketsource
 :   Serva
MOPS
  SERA29836.03
 :  Serva
Marketsource
 :  1132-61-2
MOPS
Glass bottle for solids, 500 g
 :  1132-61-2
1.060,40 kr
1 * 500 g
 :  3-Morpholinopropane-1-sulphonic acid, 3-(N-Morpholino)propanesulphonic acid
  •  :1132-61-2
  •  :C₇H₁₅NO₄S
  •  :MFCD00006183
  •  :209.27 g/mol
  •  :13,06288
  •  :> 250 °C
  •  :110 °C (closed cup)
  •  :Stue temperatur
  •  :214-478-5
  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g