Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
MES monohydrat

MES monohydrat

Glass bottle for solids, 500 g
Catalog # SERA29834.04
Marketsource
Supplier:   Serva
MES monohydrat
Catalog # SERA29834.04
Supplier:  Serva
Marketsource
CAS Number:  145224-94-8
MES monohydrat

Glass bottle for solids, 500 g

CAS Number:  145224-94-8
Price
1.508,46 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Synonymer:  MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
  • CAS nummer:145224-94-8
  • Formel:C₆H₁₃NO₄S·1H₂O
  • MDL nummer:MFCD00149409
  • Molecular Weight:213.25 g/mol
  • Merck Index:13,05929
  • Smeltende Pt:> 300 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:224-632-3
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer