Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
MES monohydrat

MES monohydrat

Glass bottle for solids, 1 kg
Catalog # SERA29834.03
Marketsource
Supplier:   Serva
MES monohydrat
Catalog # SERA29834.03
Supplier:  Serva
Marketsource
CAS Number:  145224-94-8
MES monohydrat

Glass bottle for solids, 1 kg

CAS Number:  145224-94-8
Price
2.710,75 kr
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
  • CAS Number:145224-94-8
  • Formula:C₆H₁₃NO₄S·1H₂O
  • MDL Number:MFCD00149409
  • Molecular Weight:213.25 g/mol
  • Merck Index:13,05929
  • Melting Pt:> 300 °C
  • Storage Temperature:Stue temperatur
  • EINECS:224-632-3
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.