MES monohydrat
Glass bottle for solids, 100 g
Katalog # SERA29834.02
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
- CAS nummer:145224-94-8
- Formel:C₆H₁₃NO₄S·1H₂O
- MDL nummer:MFCD00149409
- Molecular Weight:213.25 g/mol
- Merck Index:13,05929
- Smeltende Pt:> 300 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:224-632-3
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g



