Celite® 545, Supelco®
Fibre carton, 10 kg
: Merck
: DE, Diatomaceous earth, flux-calcined
- :68855-54-9
- :SiO₂
- :MFCD00132803
- :60.08 g/mol
- :M-Clarity™ quality level = MQ100
- :272-489-0
- Solubility:- 0.001 g/l (20 °C)
- Bulk Density:300 kg/m3
- pH:10 (100 g/l, H2O, 20 °C)
- Pakning:Fibre carton
- Pakn:10 kg



