Celite® 545, Supelco®
Plastic bottle, 250 g
: Merck
: DE, Diatomaceous earth, flux-calcined
- :68855-54-9
- :SiO₂
- :MFCD00132803
- :60.08 g/mol
- :M-Clarity™ quality level = MQ100
- :272-489-0
Documents
- Solubility:- 0.001 g/l (20 °C)
- Bulk Density:300 kg/m3
- pH:10 (100 g/l, H2O, 20 °C)
- Pakning:Plastic bottle
- Pakn:250 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



