Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Methylenblåt

Methylenblåt

Glass bottle for solids, 25 g
Katalog # SERA29198.01
Irriterende
Marketsource
Leverandør:   Serva
Methylenblåt
Katalog # SERA29198.01
Leverandør:  Serva
Marketsource
CAS nummer:  61-73-4
Methylenblåt

Glass bottle for solids, 25 g

CAS nummer:  61-73-4
Price
216,56 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  Basic Blue 9, Methylthioninium chloride, Methylene blue chloride, 3,7-Bis(dimethylamino)phenothiazin-5-ium chloride, Swiss Blue
  • CAS nummer:61-73-4
  • Formel:C₁₆H₁₈ClN₃S
  • MDL nummer:MFCD00150008
  • Molecular Weight:319.86 g/mol
  • Merck Index:12,06137
  • Smeltende Pt:180 °C
  • Tæthed:1,757 g/cm³ (22 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-515-2
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.