L-Methionin
Glass bottle for solids, 1 kg
Katalog # SERA28821.03
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
- CAS nummer:63-68-3
- Formel:C₅H₁₁NO₂S
- MDL nummer:MFCD00063097
- Molecular Weight:149.21 g/mol
- Merck Index:13,06004
- Kogende Pt:278 °C (1013 hPa)
- Smeltende Pt:136 °C
- Tæthed:1,373 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-562-9
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 kg



