Orders LinkContactUsLinkComponent
L-Methionin
Glass bottle for solids, 25 g
  SERA28821.01
Marketsource
 :   Serva
L-Methionin
  SERA28821.01
 :  Serva
Marketsource
 :  63-68-3
L-Methionin
Glass bottle for solids, 25 g
 :  63-68-3
1 * 25 g
 :  (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
  •  :63-68-3
  •  :C₅H₁₁NO₂S
  •  :MFCD00063097
  •  :149.21 g/mol
  •  :13,06004
  •  :278 °C (1013 hPa)
  •  :136 °C
  •  :1,373 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :200-562-9
  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g