L-Methionin
Glass bottle for solids, 25 g
SERA28821.01
Marketsource
: Serva
: (S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
- :63-68-3
- :C₅H₁₁NO₂S
- :MFCD00063097
- :149.21 g/mol
- :13,06004
- :278 °C (1013 hPa)
- :136 °C
- :1,373 g/cm³ (20 °C)
- :Stue temperatur
- :200-562-9
- Pakning:Glass bottle for solids
- Pakn:25 g