L(+)-Leucin
Glass bottle for solids, 500 g
Catalog # SERA27690.03
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine
- CAS Number:61-90-5
- Formula:C₆H₁₃NO₂
- MDL Number:MFCD00002617
- Molecular Weight:131.17 g/mol
- Merck Index:14,05451
- Melting Pt:300 °C
- Density:1,165 g/cm³ (20 °C)
- Storage Temperature:Stue temperatur
- EINECS:200-522-0
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:500 g



