Orders LinkContactUsLinkComponent
L(+)-Leucin
L(+)-Leucin
Glass bottle for solids, 100 g
  SERA27690.02
Marketsource
 :   Serva
L(+)-Leucin
  SERA27690.02
 :  Serva
Marketsource
 :  61-90-5
L(+)-Leucin
Glass bottle for solids, 100 g
 :  61-90-5
1 * 100 g
 :  (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine
  •  :61-90-5
  •  :C₆H₁₃NO₂
  •  :MFCD00002617
  •  :131.17 g/mol
  •  :14,05451
  •  :300 °C
  •  :1,165 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :200-522-0
  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g