L(+)-Leucin
Glass bottle for solids, 100 g
SERA27690.02
Marketsource
: Serva
: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine
- :61-90-5
- :C₆H₁₃NO₂
- :MFCD00002617
- :131.17 g/mol
- :14,05451
- :300 °C
- :1,165 g/cm³ (20 °C)
- :Stue temperatur
- :200-522-0
- Pakning:Glass bottle for solids
- Pakn:100 g