Cellulose for preparative chromatography
Cellulose micro-crystalline Avicel®, 10 - 160 µm, 25 kg
Micro-crystalline cellulose is a hydrophilic polysaccharide packing material preferred for the raw separation of amino acids and related compounds. Cellulose is also used for the gentle purification of bio-molecules. Because of the organic nature of cellulose, it can only be used in pre-swollen state and under gentle or hydrostatic pressure.
- Partikelstørrelse:10 - 160 µm
- Pakn:25 kg
- Beskrivelse:Cellulose micro-crystalline Avicel®



