Cellulose for preparative chromatography
Cellulose micro-crystalline Avicel®, 10 - 160 µm, 2,5 kg
Supplier: MERCK PRODUCTION CHEMICALS
Cellulose for preparative chromatography
Catalog # 1.02331.2500
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Jump to:
Product Details & Documents
Micro-crystalline cellulose is a hydrophilic polysaccharide packing material preferred for the raw separation of amino acids and related compounds. Cellulose is also used for the gentle purification of bio-molecules. Because of the organic nature of cellulose, it can only be used in pre-swollen state and under gentle or hydrostatic pressure.
Documents
Specifications
Specifications
- Partikelstørrelse:10 - 160 µm
- Pakn:2,5 kg
- Beskrivelse:Cellulose micro-crystalline Avicel®
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



