Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Iodacetamid

Iodacetamid

Glass bottle for solids, 25 g
Katalog # SERA26710.02
Irriterende
Marketsource
Leverandør:   Serva
Iodacetamid
Katalog # SERA26710.02
Leverandør:  Serva
Marketsource
CAS nummer:  144-48-9
Iodacetamid

Glass bottle for solids, 25 g

CAS nummer:  144-48-9
Price
485,40 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents
Synonymer:  2-Iodoacetamide
  • CAS nummer:144-48-9
  • Formel:ICH₂CONH₂
  • MDL nummer:MFCD00008028
  • Molecular Weight:184.96 g/mol
  • Kogende Pt:266 °C (1013 hPa)
  • Smeltende Pt:91…94 °C
  • Opbevaringstemperatur:Køleskab
  • EINECS:205-630-1
  • FN:2811
  • ADR:6.1,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.