8-Quinolinol
Glass bottle for solids, 100 g
SERA25368.02
Marketsource
: Serva
: 8-Hydroxyquinoline
- :148-24-3
- :C₉H₇NO
- :MFCD00006807
- :145.16 g/mol
- :13,04869
- :267 °C (1013 hPa)
- :71,2 °C
- :1,03 g/cm³ (20 °C)
- :Stue temperatur
- :205-711-1
- Pakning:Glass bottle for solids
- Pakn:100 g