Orders LinkContactUsLinkComponent
8-Quinolinol
Glass bottle for solids, 100 g
  SERA25368.02
Marketsource
 :   Serva
8-Quinolinol
  SERA25368.02
 :  Serva
Marketsource
 :  148-24-3
8-Quinolinol
8-Quinolinol
Glass bottle for solids, 100 g
 :  148-24-3
1 * 100 g
 :  8-Hydroxyquinoline
  •  :148-24-3
  •  :C₉H₇NO
  •  :MFCD00006807
  •  :145.16 g/mol
  •  :13,04869
  •  :267 °C (1013 hPa)
  •  :71,2 °C
  •  :1,03 g/cm³ (20 °C)
  •  :Stue temperatur
  •  :205-711-1
  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g